Learn More
Fisher Science Education™ Gram Stain Kit
SCI_ED
Catalog No. S25344 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
S25344 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. S25344
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
  • Three large-volume pipets
  • Instructions
Bacteria stain set for Gram negative and positive results.
  • Uses 1% Gentian violet solution, Gram's iodine solution, 95% ethanol (decolorizer), and 1% safranin solution
  • Gram's iodine solution packaged in PVC bottle
  • Maximum performance over 12-month shelf life ensured
  • This product is an Educational Grade Chemical — no C of A is available
Specifications
Quantity 1 Kit
Includes Three large volume pipets, instructions, 250mL 1% Gentian Violet solution, 250mL Gram's Iodine solution, 250mL decolorizer, 250mL safranin solution, 250mL 95% ethanol
Format Bottle
Stain Type Gram Stain
Product Type Gram Stain Kit