Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Hazard and NFR Identification Write-On Labels
GREENER_CHOICESDPSCI_ED

Catalog No. S43188 Shop All Fisherbrand Products
Change view
Click to view available options
Width (English):
3.88 in.
5 in.
Height (English):
1.5 in.
3 in.
Height (Metric):
3.81 cm
7.6 cm
Quantity:
100/Roll
1000/Roll
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Height (English) Height (Metric) Quantity
S43188 3.88 in. 1.5 in. 3.81 cm 1000/Roll
S475646 5 in. 3 in. 7.6 cm 100/Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S43188
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.

Blank hazard classification diamond can be filled in with the appropriate ratings.

Large Labels

  • Spaces for Chemical Name, Hazards and Precautions
  • Check boxes for goggles, gloves, apron and fume hood
  • Dimensions: 5W × 3 in.H (12.7 × 7.6cm)
Small NFR Labels
  • Spaces for Chemical Name, Owner and Date
  • Dimensions: 3.88W × 1.5 in.H (9.9 × 3.8cm)

Specifications

Format Label
Product Type Hazard Identification Label
For Use With (Application) Chemical hazard
Quantity 1000/Roll
Width (English) 3.88 in.
Height (English) 1.5 in.
Height (Metric) 3.81 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.