Learn More
Fisher Science Education™ Replacement Tubular Bulb
SCI_ED
Catalog No. S74021
Change view
Click to view available options
For Use With (Equipment):
Non LED model microscopes
Catalog No. For Use With (Equipment)
S74021 Non LED model microscopes
1 options
Catalog No. S74021 Supplier Fisher Science Education™ Supplier No. 800400
Add to Cart
Add to Cart
Replacement Tubular Bulb for stereoscopic microscopes.
  • 12V, 10W tubular replacement bulb for National Optical 405 Series, 446 Series, 450 series, and DC4-456H
Specifications
For Use With (Equipment) Non LED model microscopes
Voltage 12 V
Wattage 10 W
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov