Learn More
Fisher Science Education™ Replacement LED Bulb
SCI_ED
Catalog No. S74023
Change view
Click to view available options
For Use With (Equipment):
450 LED Series Stereoscopic Microscopes
Catalog No. For Use With (Equipment)
S74023 450 LED Series Stereoscopic Microscopes
1 options
Catalog No. S74023 Supplier Fisher Science Education™ Supplier No. 800453
Add to Cart
Add to Cart
Replacement LED Bulb for stereoscopic microscopes.
  • Replacement LED bottom bulb for National Optical 452 and 453 LED models
Specifications
Type Replacement Bulb
For Use With (Equipment) 450 LED Series Stereoscopic Microscopes
Wattage 10 W
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov