Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Pencils
SCI_ED

Catalog No. S04194 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Multi
Quantity:
12 Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
S04194 Multi 12 Pk.
S04191 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04194
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used in the science classroom.

Specifications

Product Type Pencil
Color Multi
For Use With (Application) Used in the science classroom
Quantity 12 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.