Learn More
Fisher Science Education™ Repeating Pipet Filler
SCI_ED
Catalog No. S32250 Shop All Fisherbrand Products
Change view
Click to view available options
Pipette Filler w/plunger; For 10mL pipets; 6/Pk.
Catalog No.
S32250 Pipette Filler w/plunger; For 10mL pipets; 6/Pk.
1 options
This item is not returnable. View return policy
Catalog No. S32250
This item is not returnable. View return policy
Fits pipets up to 10mL.
Repeating pipet filler has a molded PVC body with rubber bellows. The control top adjusts the plunger stroke for repeating manually preset volume up to 10mL.
  • Depress plunger to expel air
  • Release plunger to draw up liquid
  • Depress plunger to release liquid
Specifications
Autoclavable Not Autoclavable
Material PVC, Rubber
Type Repeating Pipette Filler
For Use With (Application) Pipets up to 10mL
For Use With (Equipment) 10mL
Includes Plunger