Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Science Superhero Magnetic Dry Erase Board
SCI_ED

Catalog No. FSEBOARD Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
12 x 18 in. (30.5 x 45.7 cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
FSEBOARD 12 x 18 in. (30.5 x 45.7 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FSEBOARD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Pen and clips

Featuring the STEMcredibles superheroes.

  • Framed
  • Magnetic
  • 12 x 18 in.

Specifications

Includes Pen and clip
Dimensions (L x W) 12 x 18 in. (30.5 x 45.7 cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.