Learn More
Fisher Science Education™ Total Alkalinity Test Kit
SCI_ED
Catalog No. S01955 Shop All Fisherbrand Products
Change view
Click to view available options
Total Alkalinity Test Kit
Catalog No.
S01955 Total Alkalinity Test Kit
1 options
This item is not returnable. View return policy
Catalog No. S01955
This item is not returnable. View return policy
Includes
Total alkalinity indicator, 30mL; alkalinity titrant, 60mL; plastic vial
Use this test kit for analysis of natural water samples using a titration test method.
Alkalinity values of 20 to 200ppm are common in freshwater ecosystems and those below 10ppm indicate poorly buffered streams.
  • One drop equals 10ppm alkalinity as CaCO3
  • Perfom the test and compare the color change to the EndPoint ID™ photograph on the instructions
Specifications
For Use With (Application) Use this test kit for analysis of natural water samples using a titration test method
Includes 30ml total alkalinity indicator, 60ml alkalinity titrant, plastic vial
Product Type Total Alkalinity Test Kit