Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Water Quality Saddle Bag
SCI_ED

Catalog No. S02645 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Application):
For the study of water pollution and its effects
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S02645 For the study of water pollution and its effects
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Choking hazard (1) Small parts. Not for children under 3 yrs.

Catalog No. S02645
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Step-by-step laminated picture instructions, an instructor's manual with background information, reproducible student data sheets and color-coded reagents

For study of water pollution and its effects.

  • Designed to wear on the side of the individual for safety and ease
  • Contains: alkalinity P/T/acidity, alkalinity=1 drop=10ppm/25mL, acidity =0 to 500ppm, hardness (total), 1 drop =2ppm/25mL, dissolved oxygen (0 to 10ppm), nitrate (0 to 50ppm), and pack of 100 pH test strips (2 to 9 range)

Order Info

Each test kit has enough materials to perform over 100 tests and can be used by a group of 30 students or more.

Specifications

Product Type Water Quality Saddle Bag
Includes Step-by-step picture instructions and color coded reagents
For Use With (Application) For the study of water pollution and its effects
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.