Learn More
Fisher Science Education™ 160-Series University/Laboratory Compound Microscopes
SCI_ED
Catalog No. S23876
Change view
Click to view available options
Objective Types:
Semi Plan
Catalog No. Objective Types
S23876 Semi Plan
1 options
Catalog No. S23876 Supplier Fisher Science Education™ Supplier No. 168SP
Add to Cart
Add to Cart
Designed for university and laboratory use. Fisherbrand™ 160-Series University/Laboratory Compound Microscope has good optical clarity and reliable mechanical engineering assure years of top performance.
  • Remarkable optical clarity
  • Choice of objective lenses:  ASC, semi-plan, plan and phase
  • LED energy-efficient illumination 
  • Large, built-in, low-profile, rackless stage
  • Coaxial focusing
  • Magnification:  40X, 100X, 400X, and 1000X
Specifications
Condenser Type Abbe 1.25 N.A
Head Style Binocular
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Objective Types Semi Plan
Product Type Microscope
Warranty Limited lifetime, excluding bulb, cord and fuse
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov