Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuFet™ Solid-State pH/ATC Combination Electrodes - Mercury - Free

Catalog No. 13620758 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
DIN
Mini-DIN
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-620-758 Mini-DIN
13-620-755 DIN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-620-758

missing translation for 'validMainframeMsg'

Rugged, unbreakable ISFET sensor for difficult measurements

ISFET solid-state pH sensor plus reference and ATC probe—in one tough epoxy body

  • Use directly with accumet™ AR, AB15, AP15P or BASIC
  • Replaces fragile glass pH-sensing membrane with rugged, solid-state Ion-Selective Field Effect Transistor (ISFET)
  • Reference cell contains gelled Ag/AgCl electrolyte, for high performance with low maintenance
  • Porous junction provides extra-long service
  • Built-in temperature probe for accurate measurements in a minimum of space
  • Durable epoxy body takes all the rough handling you can dish out
  • Nonglass sensor remains unscathed, even by abrasive soil samples encountered in environmental testing
  • Gives you high reproducibility, excellent stability and fast response time without risk of breaking a glass body or delicate glass bulb
A cost-effective solution for high-risk applications
  • For aqueous solutions, viscous samples, even semisolids
  • For situations where breakable glass electrodes could be hazardous—food, beverage and pharmaceutical processing, for example
  • Ideal both for rapid, routine, high-workload measurements and for reliable measurement of difficult samples in environmental, pharmaceutical, food and beverage applications
  • Rapid, reproducible results even in thick, glutinous samples: simply wipe unbreakable sensing element to remove sticky, viscous substances—even peanut butter and bread dough—to eliminate sample carryover

For use with a variety of meters

  • accuFet electrode 13-620-755 with DIN connector is for use with accumet Research (AR) and accumet Basic (AB) benchtop models—no adapter needed
  • accuFet electrode 13-620-758 with mini-DIN connector is for use with discontinued BASIC series meters directly
  • accuFet electrode 13-620-AP20 with waterproof Twist-lock connector is for use with discontinued accumet portable meters AP25 and AP15

TRUSTED_SUSTAINABILITY

Specifications

Connector Types Mini-DIN
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type None
For Use With (Application) Beverage, Food, Pharmaceutical
For Use With (Equipment) Basic benchtop meter
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.