Learn More
Fisherbrand™ accuflex™ Electrode Support
Catalog No. 13637671 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
Electrode support arm bracket for meter
Catalog No. Includes
13-637-671 Electrode support arm bracket for meter
1 options
Catalog No. 13-637-671
Add to Cart
Add to Cart
For accumet™ benchtop meters
  • Attaches to meter base with bracket supplied
  • Holds one micro electrode and three standard electrodes
  • Permits full up/down and left/right positioning of electrodes
  • Made of chemical-resistant plastic
TRUSTED_SUSTAINABILITY
Specifications
Type Support Arm and Bracket
For Use With (Application) pH
Includes Electrode support arm bracket for meter
Material Plastic