Learn More
Fisherbrand™ accumet™ AC Adapters
Catalog No. 13636100 Shop All Fisherbrand Products
Change view
Click to view available options
For Accumet AP60, AP100, AR, and AB series meters; 110/220V
Catalog No.
13-636-100 For Accumet AP60, AP100, AR, and AB series meters; 110/220V
1 options
Catalog No. 13-636-100
AC adapter features a 3-prong grounding plug.
This power supply fits the following accumet instruments: AB15, AB15P, AB30, AB40, AP61, AP62, AP63, AP110, AP115, AP125. AR15, AR20, AR25, AR40, AR60.
TRUSTED_SUSTAINABILITY
Specifications
Type AC Adapter
Electrical Requirements 110/220 V, 60 Hz
For Use With (Application) pH
Frequency 60 Hz
Includes 3-prong grounding plug
Voltage 110/220 V
For Use With (Equipment) Accumet instruments: AB15, AB15P, AB30, AB40, AP61, AP62, AP63, AP110, AP115, AP125, AR15, AR20, AR25, AR40, AR60