Learn More
Fisherbrand™ accumet™ AP65 Portable Conductivity Meter
Catalog No. 13636AP65A Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Portable Conductivity Meter
Portable Conductivity Meter Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13636AP65A Portable Conductivity Meter
13-636-AP65 Portable Conductivity Meter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13636AP65A
Includes
  • Meter Kit includes: Meter, 9V battery, 2-cell probe with ATC built in, 1.0cm-1 K constant, and operation manual in a handy, hard carrying case.
  • Meter Only includes Meter, 9V battery and operation manual.
Use inside the lab and on the job site
Measures conductivity in μS/cm, resistivity in megohm·cm, TDS in ppm, and °C. Trim shape and rubberized wraparound guards allow easy transportation inside a shirt pocket and comfortable, single-handed operation. Tough ABS plastic housing stands up to harsh usage and is waterproof to IP67 standards. Meter even floats if dropped in water.
  • Big, readable display: LCD (1.9375W × 2.5 in.H [4.9 × 6.4cm]) shows measurements in large 0.4375 in.H (1.1cm) characters; other characters are 0.75 in.H (0.6cm)
  • Features six sealed, soft-touch entry keys as well as plain-language on-screen prompts and error messages that allow easy navigation through operation menus
  • Integral ATC; accepts 2- and 4-cell conductivity probes (available separately)
Order Info
Dimensions: 7.5L x 2.875W x 1.375 in. H (19 x 7.3 x 3.5cm). Net weight: 0.7 lb. (0.32kg).
Warranty and Services
Two years
TRUSTED_SUSTAINABILITY
Specifications
Includes Meter, 9V battery and operation manual
Type Portable Conductivity Meter