Learn More
Fisherbrand™ accumet™ AP65 Portable Conductivity Meter
Catalog No. p-186207 Shop All Fisherbrand Products
Click to view available options
Type:
Portable Conductivity Meter
Portable Conductivity Meter Kit
Includes
  • Meter Kit includes: Meter, 9V battery, 2-cell probe with ATC built in, 1.0cm-1 K constant, and operation manual in a handy, hard carrying case.
  • Meter Only includes Meter, 9V battery and operation manual.
Use inside the lab and on the job site
Measures conductivity in μS/cm, resistivity in megohm·cm, TDS in ppm, and °C. Trim shape and rubberized wraparound guards allow easy transportation inside a shirt pocket and comfortable, single-handed operation. Tough ABS plastic housing stands up to harsh usage and is waterproof to IP67 standards. Meter even floats if dropped in water.
  • Big, readable display: LCD (1.9375W × 2.5 in.H [4.9 × 6.4cm]) shows measurements in large 0.4375 in.H (1.1cm) characters; other characters are 0.75 in.H (0.6cm)
  • Features six sealed, soft-touch entry keys as well as plain-language on-screen prompts and error messages that allow easy navigation through operation menus
  • Integral ATC; accepts 2- and 4-cell conductivity probes (available separately)
Order Info
Dimensions: 7.5L x 2.875W x 1.375 in. H (19 x 7.3 x 3.5cm). Net weight: 0.7 lb. (0.32kg).
Warranty and Services
Two years
TRUSTED_SUSTAINABILITY
Specifications
Includes Meter, 9V battery, 2-cell probe with ATC built in, 1.0cm-1 K constant, and operation manual in a handy, hard carrying case
Type Portable Conductivity Meter Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.