Learn More
Fisherbrand™ accumet™ Membrane Caps for Gas-Sensing Combination ISEs
Catalog No. 13620512 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
12 mm
Catalog No. Diameter (Metric)
13-620-512 12 mm
1 options
Catalog No. 13-620-512
Add to Cart
Add to Cart
Set of three replacement membrane caps with O-rings.
  • For Ammonia Gas-Sensing Combination ISEs 13-620-508, 13-620-509; Carbon Dioxide Gas-Sensing Combination ISEs 13-620-510, 13-620-511
  • Set of three replacement membrane caps
  • With O-rings
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ammonia Gas-sensing Combination ISE
Type Membrane Cap
Diameter (Metric) 12 mm