Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ Micro Glass Mercury-Free Combination Electrodes

Catalog No. p-4950175 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
5 in.
10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
13-620-851 10 in.
13-620-850 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-620-851
Only null left
Add to Cart
Add to Cart

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

  • Glass body (3 x 38mm) with 3mm teflon stem
  • Refillable
  • TRIS compatible
  • Mercury-free
  • Double junction
  • pH Range: 0 to 14

TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Junction Double
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
For Use With (Application) Capillary Tubes, Cuvettes, Microwell Plates, Micro Test Tubes, NMR Tubes, Serum Cups
Temperature (English) Operating 32°F to 176°F
Length (English) 10 in.
Length (Metric) 25.4 cm
Length (Metric) Stem 15.2 cm
Diameter (English) 0.12 in.
Diameter (Metric) 3 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.