Learn More
Fisherbrand™ accumet™ Research AR40 Benchtop Dissolved Oxygen Meter Kit
Catalog No. 13636AR40 Shop All Fisherbrand Products
Change view
Click to view available options
AR40 benchtop dissolved oxygen meter kit
Catalog No.
13-636-AR40 AR40 benchtop dissolved oxygen meter kit
1 options
Catalog No. 13-636-AR40
Includes
meter, power supply, accumet Self-Stirring BOD Probe (13-620-SSP), Membrane Kit (13-637-DOM), and manual.
Easy-to-use dissolved oxygen meter provides accuracy, reliability and intuitive operation.
Easy-to-use dissolved oxygen meter provides accuracy, reliability and intuitive operation
  • Extra-large 640 × 480 LCD offers touchscreen control
  • Configurable display shows measurements in 3/4 in.H (2cm) figures
  • Context-specific interface, extensive menu options and help screens make operation a breeze
  • Internal diagnostics routines help assure reliable measurements
Extensive data storage and output options
  • Features 250 data-point memory
  • Bidirectional RS-232-C outout sends data to printer or computer
  • Programmable data storage; allows output when reading is stable, at timed interval, or on demand
  • Allows printout interval of 1 to 9999 seconds
  • Programmable alarm
Automatic calculations
  • Calculates five-day BOD results, OUR, SOUR
  • Provides automatic barometric pressure and salinity compensation
Order Info
Call your Fisher Scientific Customer Service Representative for full details.
Warranty and Services
accumet Three-year Warranty. accumet Model AR40 is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.