Learn More
Fisherbrand™ Adjustable Stand for Model 705 Sonic Dismembrator
SDP
Catalog No. FB4438 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Model 705 sonic dismembrator
Catalog No. For Use With (Equipment)
FB4438 Model 705 sonic dismembrator
1 options
Catalog No. FB4438
Heavy duty and vertically adjustable
This Fisher Scientific™ Adjustable Stand for the Model 705 Sonic Dismembrator keeps horn level and allows adjustment of horn in / out of the microplate. Heavy duty and vertically adjustable.
For 4611 96-tip horn.
Specifications
Product Type Adjustable Stand
For Use With (Application) Ideal for allows adjustment of horn in, out of there microplate
For Use With (Equipment) Model 705 sonic dismembrator