Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cheesecloth Mini-Wipes
SCI_ED

Catalog No. S04825 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
17.8 x 35.5 cm (7 x 14 in.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S04825 17.8 x 35.5 cm (7 x 14 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S04825
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cheesecloth Mini-Wipes made of certified 100% reagent-grade cotton.

  • Each 4 in. square pad (8-ply) unfolds to approximately 7 x 14 in.
  • Pure cotton with grade 10 (mesh size)
  • Bleached
  • Meets all lab requirements for cleanliness, softness, absorbency, and strength
  • Can be used on microscopes, optical surfaces, glassware, pH electrodes and will not scratch sensitive surfaces
  • Absorbs 6 times their own weight of both solvents and aqueous solutions
  • Individually dispensed wipes eliminate time-consuming cutting of bolts of cloth and dust, dirt, and waste associated with handling difficult-to-store rolls

Specifications

Product Type Cheesecloth Miniwipe
Length (English) 7 in.
Length (Metric) 18 cm
Width (English) 14 in.
Width (Metric) 36 cm
Material Cotton
Dimensions (L x W) 17.8 x 35.5 cm (7 x 14 in.)
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.