Learn More
Fisherbrand™ CO2 Backup Systems
Catalog No. 13990113 Shop All Fisherbrand Products
Change view
Click to view available options
Factory-installed
Catalog No.
13-990-113 Factory-installed
1 options
Catalog No. 13-990-113
Requires
CO2 cylinder
For Fisher Scientific Isotemp™ Basic Freezers
Freestanding and factory-installed systems inject liquid CO2 into the cabinet when cabinet temperature warms to preset level.
  • Adjustable in 1°C increments to -75°C (-103° F)
  • Digital display constantly indicates actual cabinet temperature
  • Liquid flow is shut off automatically when freezer door or lid is opened
  • Indicator lights for system activation, low battery, empty cylinder
  • Powered by rechargeable battery
Order Info
Factory-installed model must be specified when ordering freezer. For freestanding model, specify freezer voltage when ordering.
TRUSTED_SUSTAINABILITY
Specifications
Type Backup, CO2
Temperature Range 1°C to 75°C
For Use With (Equipment) Isotemp Basic -86 deg. Freezers
Model Factory-installed