Learn More
Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Catalog No. p-4017676 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
US Standard/Pin Plug to BNC
For Use With (Application):
dapts electrode with U.S. Standard/Pin plugs to BNC input jack
Catalog No. Connector Types For Use With (Application)
13-620-498 US Standard/Pin Plug to BNC dapts electrode with U.S. Standard/Pin plugs to BNC input jack
1 options
Catalog No. 13-620-498
Only null left
Add to Cart
Add to Cart
Adapters connect electrodes to various input jacks and are mercury free
  • Adapters connect electrodes to various input jacks
TRUSTED_SUSTAINABILITY
Specifications
Connector Types US Standard/Pin Plug to BNC
For Use With (Equipment) Electrode
Type Connection Adapter
For Use With (Application) dapts electrode with U.S. Standard/Pin plugs to BNC input jack