Learn More
Fisherbrand™ Extension Support for Spherical and Hemispherical Mantles
Catalog No. 1147242 Shop All Fisherbrand Products
Change view
Click to view available options
250mL
Catalog No.
11-472-42 250mL
1 options
Catalog No. 11-472-42
For attaching Spherical Mantles and Hemispherical Mantle-Bottoms (see above) to lattice. Consists of steel ring with heat-resistant strap basket.