Learn More
Fisherbrand™ FH120DH Dispensing Handle
Catalog No. 13310667 Shop All Fisherbrand Products
Click to view available options
For Use With:
FH100D or FH100DX
Catalog No. 13-310-667
Offers contamination-free operation
Ideal for automated process applications.
Ease of use
  • Contamination free operation
  • Adaptability with various pump models and tubing sizes
  • Intuitive controls -- Start/Stop, Dispense/Purge and Prime
Ease of maintenance
  • IP31 enclosure rating allows for simple wipe down clean up
  • Tubing runs the full length of the handle -- no interior surfaces to clean
Diverse performance range
  • Accommodates a wide range of tubing sizes from 0.8 to 8.0mm ID
  • Dispense volumes from micro liters to more than 5 L/min., depending on the tubing size
Ergonomic Design
  • Easy-grip design for use with glove-protected hands
  • Tilted head permits user to maintain a natural hand position thereby preventing hand injury
  • Momentary switch helps reduce hand fatigue by eliminating the need for constant pressure on the switch
Warranty and Services
None
Specifications
Description Dispensing handle
For Use With FH100D or FH100DX
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.