Learn More
Fisherbrand™ Isotemp™ Chiller Installation Kit
Catalog No. 13265235 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Installation Kit
Catalog No. Type
13-265-235 Installation Kit
1 options
Catalog No. 13-265-235
Installation kit
For use with Fisher Scientific Isotemp Recirculating Chillers
TRUSTED_SUSTAINABILITY
Specifications
Type Installation Kit
Includes 0.5 x 0.5 in. MPT fittings, 25 ft. of 0.62 in. I.D. Polybraid hose, hose clamps and hose insulation