Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cover for Precision™ Water Baths, 4 holes, for 10L Water Bath

Catalog No. 1546230Q Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
15-462-30Q 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-462-30Q
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Fisherbrand™ Covers are for use with Precision Dubnoff and Shallow-Form Shaking Water Baths.

Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
TRUSTED_SUSTAINABILITY

Specifications

Description Concentric Ring Cover 4 Holes
Capacity (Metric) 10 L
For Use With (Equipment) Precision 10L General Purpose Water Baths
Material Stainless Steel
No. of Positions 4
Warranty 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.