Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Enzyme Storage Freezers

Catalog No. 13986154ES Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
5 cu. ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
13-986-154ES 5 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-986-154ES

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Includes

Two cases of freezer packs to blanket samples and protect from temperature oscilliations that occur when main door is open.

Hard-working freezers specifically designed to meet enzyme storage needs

  • Choice of 5 or 20.6 cu. ft. capacity
  • Temperature range: -20° to -23°C
  • Average temperature stability: ±4°C (air), ±2°C with freezer packs
  • Audible and visual alarm if temperature rises above -20°C
  • Manual defrost for sensitive sample storage
  • One hinged lockable door
  • Durable construction designed to withstand rigorous use
  • White epoxy-coated steel interior with white, heavy, baked-on polyester enamel finish exterior

Order Info

For 115V 60Hz.

Certifications

13-986-154ES is UL/cUL listed

Warranty and Services

13 months parts and labor, 5 years on compressors

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 5 cu. ft.
Capacity (Metric) 142 L
Style Undercounter
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.