Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Explosion-Proof Freezers

Catalog No. 13986150 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
583.3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-986-150 583.3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-986-150

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Includes

4 fixed Interior shelves, 1 Wire Basket, 6 door shelves

For storing volatile materials in areas with explosive atmospheres or in hazardous locations

  • Temperature range: -20° to -12°C
  • Adjust setpoint while running
  • Capacities: 20 cu. ft. (566.34L)
  • Manual defrost
Safety Features
  • Red warning label on front: “Explosion-Proof Freezer” Compressors have enclosed overloads, relays
  • Top-mounted, explosion-proof thermostats
  • Static hot-wall condensers and evaporators are free of electric fans
Construction
  • White, baked-on enameled steel exteriors
  • Hinged solid door
  • White enameled steel interiors
  • R134A, CFC Free refrigerant, polyurethane foam insulation

Order Info

Not equipped with line cord and plug; must be hardwired into an appropriately rated conduit box to comply with NEC code.

Service Plan

Service agreement available.

Certifications

UL listed meets NFPA 45 and 70

Warranty and Services

13 months, parts and labor

TRUSTED_SUSTAINABILITY

Specifications

Type Explosion-Proof Freezer
Capacity (English) 20.6 cu. ft.
Capacity (Metric) 583.3 L
Temperature Range -12°C to -20°C
Door Style Hinged
No. of Doors 1 (1 compartment)
Hinge Side Right
Insulation Polyurethane Foam
Interior Epoxy Coated Steel
Exterior Finish Epoxy Coated Steel
Shelves 4 Fixed
Door Shelves 6 Fixed
Certifications/Compliance cCSAus
Casters Adjustable Leveling Feet
Color White
Control Type Mechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
Warranty 13 Months Parts and Labor
Defrost Manual
Model 20EFEEFSA
Dimensions (D x W x H) Interior 21 x 27 x 57.5 in. (53.34 x 68.58 x 146.05 cm)
Dimensions (L x W x H) Exterior 39.61 x 33 x 67.25 in. (100.6 x 83.8 x 170.8 cm)
Access Security Door Lock
Amperage 5 A
Plug Type Not Provided
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Wattage 575 w
Compressor Bottom-mounted
Refrigerant R134a, 100% CFC Free
Shipping Weight (English) 230 lb.
Shipping Weight (Metric) 104.3 kg
Weight (English) 200 lb.
Weight (Metric) 90.7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.