Learn More
Fisherbrand™ Isotemp™ LN2 Backup Systems for Ultra-Low Temperature Freezers
Catalog No. 1950530 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Factory-installed
Field-installed
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
1950530 Field-installed
1950528 Factory-installed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1950530
For use with Isotemp Upright and Chest Ultra-Low Temperature Freezers
LN2 Backup System: Maintains temperature down to -85°C
TRUSTED_SUSTAINABILITY
Specifications
Type Field-installed
For Use With (Equipment) Isotemp™ Ultra low temperature freezer