Learn More
Fisher Scientific™ Level C Responder Kits
Catalog No. BE10KITLL
Change view
Click to view available options
Size:
Large
Catalog No. Size
BE10KITLL Large
1 options
Catalog No. BE10KITLL
Includes
  • Liithium battery-powered 3M™ Breathe Easy™ (BE10) PAPR
  • One DuPont™ Tychem™ F Coverall (Large or X-Large)
  • One roll of Kappler™ Chem-Tape™
  • One pair of Large or X-Large Fisherbrand™ Nitrile Gloves
  • One pair of size 9 Ansell™ Sol-Vex™ Nitrile Gloves
  • One pair of DSC™ Haz Mat Booties
  • Sturdy, zippered nylon bag with durable carrying straps
Provides Level C protection for decon line attendants
Contains the necessary respiratory and protective apparel used by decontamination line attendants who may be assisting patients exposed to chemical or biological agents or similar contaminants for which Level C protection is required. Also suitable for other personnel as determined by competent authorities.
Kit should be donned, decontaminated, and doffed in accordance with your institution's procedures and manufacturer's guidance.
(Note: Not for reuse except and until decontaminated and inspected for reuse in accordance with your institution's procedures and manufacturer's guidance).
Order Info
  • Abbreviated descriptions for each of the components are provided here. For additional information on each component, search our web site using the component's name as the search term.
  • 3M Breathe Easy (BE10) PAPR with choice of 12-hour lithium battery pack or rechargeable 8-hour NiCd battery pack offers head and shoulder protection from a variety of chemical warfare agents and industrial chemicals. Belt-mounted blower draws air through filter and provides air to hood via flexible breathing tube.
  • PAPR includes one set of three high-efficiency 3M Cartridge FR-57 with high-efficiency filters for removing solid and liquid aerosols, and an activated/impregnated carbon layer for the removal of gases and liquid vapors. (Note: This unit is not recommended for use in atmospheres where oxygen concentration is below 19.5%; where contaminant concentrations are unknown or considered Immediately Dangerous to Life or Health [IDLH]; or where contaminant concentration exceeds the lower of applicable maximum use limitations under local or OSHA standards.)
  • For results of usage and chemical resistance testing, call 3M Occupational Health & Environmental Safety Division, Technical Services Department, at 1-800-243-4630.
  • DuPont Tychem F Coverall provides high-level splash protection against various warfare agents, as well as industrial chemicals. Low-visibility gray coverall features taped seams, elasticized face opening, ankles, and wrists. Zippered front with storm flap also utilizes double-sided tape for added protection. (Note: Tychem F is not flame-resistant; do not expose to spark, flame, extreme heat, or flammable environments).
  • For results of usage and chemical resistance testing, call DuPont Personal Protection at 1-800-931-3456 or visit www.personalprotection.dupont.com.
  • Fisherbrand Powder-Free Nitrile Exam Gloves are ideal for use as a secondary defense against chemical permeat
Specifications
Size Large