Learn More
Fisherbrand™ Micromaster™ I Brightfield Microscopes
Catalog No. 12561B Shop All Fisherbrand Products
Change view
Click to view available options
Head Style:
Binocular
Catalog No. Head Style
12-561B Binocular
1 options
Catalog No. 12-561B
Includes
Planachromatic objectives, Koehler illumination, reversed nosepiece, dust cover, immersion oil, spare fuse and spare halogen lamp
Professional quality optics at affordable prices
Ideally suited to educational and general laboratory use. Monocular, binocular and trinocular models are available.
Components
  • Eyepieces: 10X wide-field, DIN standard; binocular and trinocular models equipped with two eyepieces
  • Head: available in monocular, binocular and trinocular configurations; interchangeable and rotatable 360° with a 30° inclination; binocular and trinocular heads adjustable for interpupillary distance and dioptric differences, interpupillary adjustment range, 55 to 75mm, adapters available for photomicroscopy or videomicroscopy
  • Nosepiece: quadruple reversed nosepiece with positive click stops enabling easy slide manipulation without interference by the objectives; aids in keeping optics clean
  • Objectives: planachromatic objectives produce true color images without blurring; 4X (0.10 N.A.), 10X (0.25 N.A.), 40X (0.65 N.A. retractable), and 100X (1.25 N.A. retractable) oil immersion DIN standard objective
  • All are parfocal, parcentered and coated to resist reflection, optional objectives are available
  • Stage: precision-machined mechanical stage with oversized, low-position, coaxial control knobs; chemical-resistant finish with removable spring-clip slide holder secures slide to the stage
  • Focus control: low-position, coaxial, coarse/fine controls; adjustable safety stop prevents slide damage; tension adjustment ring permits custom user setup
  • Condenser: 1.25 N.A. Abbe condenser, fitted with an iris diaphragm and swing-in auxiliary lens for low magnification
  • Illumination: Koehler illumination with variable-intensity quartz halogen lamp, field diaphragm and collector lens with integrated drop-in filter holder; blue and frosted filters included 6V 20w lamp with solid-state dimmer and heavy-duty 3-prong cord
  • On/Off switch
  • Finish: chemical-resistant baked-on paint with stipple finish resists fingerprints and smudges
Order Info
Overall dimensions: 10.5L x 8.5W x 17 in. H (27 x 21.5 x 43cm). For 110V 50/60Hz.
Service Plan
Service agreement available.
TRUSTED_SUSTAINABILITY
Specifications
Head Style Binocular