Learn More
Fisher Scientific™ PowerGen Omni Tip™ Adapter
Catalog No. 15340150 Shop All Omni International Inc Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
15-340-150 Stainless Steel
1 options
Catalog No. 15-340-150 Supplier Fisher Scientific™ Supplier No. FA1000SB
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For Powergen homogenizers
  • Material: Stainless-steel
  • For use with OMNI TIP plastic disposable generator probes with homogenizers
Specifications
Product Type Adapter
Material Stainless Steel
For Use With (Equipment) Omni Tip™ plastic disposable generator probes