Learn More
Fisher Healthcare™ PROTOCOL™ Histobond Adhesive
Catalog No. 23245693 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
475 mL
Catalog No. Volume (Metric)
23-245693 475 mL
1 options
Catalog No. 23-245693
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Resin that enhances adhesion of specimens to slides prior to hydrating and staining
Specifications
Applications Resin that enhances adhesion of specimens to slides prior to hydrating and staining
Product Type Histobond Adhesive
Volume (Metric) 475 mL