Learn More
Fisherbrand™ Replacement Urine Hydrometer for Midget Urinometer Set (22274209)
Shop All Fisherbrand Products
Description
- Replacement urine hydrometer designed for Fisherbrand™ Midget Urinometer Set (Cat. No. 22274209)
- 0.001 specific gravity increments support precise, repeatable readings
- Accuracy: ±0.002 specific gravity for routine urinalysis workflows
- Shot-ballasted soda-lime glass construction provides stable flotation
- 110 mm length sized for use in the matching urinometer jar
Specifications
Specifications
| Type | Urine Hydrometer |
| Accuracy | ±0.002 |
| Increments | 0.001 |
| Length (Metric) | 110 mm |
| Material | Soda Lime Glass |
| For Use With (Application) | Urine analysis |