Learn More
Fisherbrand™ Rockers and Wavers: Stacking Tray
Catalog No. 02217736 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Rockers
Catalog No. For Use With (Equipment)
02-217-736 Rockers
1 options
Catalog No. 02-217-736
Add to Cart
Add to Cart
For use with Fisherbrand Incubating Rockers, Open Air Rockers and Incubating Wavers.
Stacking tray attaches to Open Air Rocker unit to add second tier, mounts 3.5 in. (8.9cm) above lower tray, includes rubber mat and all hardware
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stacking Tray
Material Aluminum
For Use With (Equipment) Rockers