Learn More
Fisherbrand™ Shorting Strap for BNC Input
Catalog No. 1362099 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Meter with BNC input jack
Catalog No. For Use With (Equipment)
13-620-99 Meter with BNC input jack
1 options
Catalog No. 13-620-99
For meters with BNC input jack.
For meters with BNC input jack
  • Connects input jacks for testing and calibration of meter
  • Metal piece and strap hangs from BNC connection when not in use; when plugged in, it provides a 0 mV signal
TRUSTED_SUSTAINABILITY
Specifications
Type Shorting Strap
For Use With (Application) pH
For Use With (Equipment) Meter with BNC input jack