Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Special-Purpose Undercounter Freezers

Catalog No. 979281 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
141.6 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
97-928-1 141.6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 97-928-1

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Compact flammable-materials storage and explosion-proof models

 

  • Adjustable temperatures from -20° to -15°C (-4° to +5°F)
  • Capacity, 5.0 cu. ft. (141.6L) Manual defrost
  • R-134a CFC-free refrigerant
  • 360w compressors
  • Baked-on, acrylic enamel exterior
  • Easy-to-clean formed interior
  • Solid door mounted to open from left

 

Flammable-Materials Storage Model

  • Accidental ignition of refrigerator contents is prevented by specially designed compressor with enclosed overload, relay, and hydraulic thermostat
  • Static condenser free of electric fan
  • Aluminum evaporator coil
  • Two fixed shelves, three door shelves
  • Requires extra clearance when recessed†
Explosion-Proof Model
  • Mechanical assembly design prevents accidental explosion of environmental vapors
  • Wiring totally enclosed in sealed pipes
  • Hydraulic thermostat enclosed by precision-machined housing
  • Two fixed shelves, three door shelves
  • Must be hardwired to an appropriately rated conduit box

 

Service Plan

Service agreement available.

Certifications

UL listed

Compliance

Flammable-Materials Storage Model complies with NFPA Standard Numbers 45 and 99 and OSHA 29CFR1910.308. Explosion-Proof Model wiring complies with NEC Articles 500-501 for use in Class 1, Groups C and D hazardous locations.

Warranty and Services

Two years parts, one year labor

TRUSTED_SUSTAINABILITY

Specifications

Type Explosion-Proof Freezer
Capacity (English) 5 cu. ft.
Capacity (Metric) 141.6 L
Temperature Range -15°C to -20°C
Insulation CFC-free
Shelves 6
Certifications/Compliance NFPA, OSHA, UL
Defrost Manual
Dimensions (D x W x H) Interior 15 x 18 x 28 in. (38.1 x 45.7 x 71.1 cm) Upper
10 x 18 x 28 in. (25.4 x 45.7 x 71.1 cm) Lower
Dimensions (L x W x H) Exterior 29.75 x 21 x 33.5 in. (74.9 x 54.6 x 85.1 cm)
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Refrigerant R134a, 100% CFC Free
Weight (English) 84 lb.
Weight (Metric) 38.1 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.