Learn More
Fisherbrand™ Squibb Urinometer Set for Urine Specific Gravity Measurement, 1.000 to 1.060 Range
Shop All Fisherbrand Products
Description
- Measures urine specific gravity from 1.000 to 1.060 for routine urinalysis
- Fine 0.001 increments support precise readings
- Accuracy: +/-0.002 for dependable results
- Includes shot-ballasted urinometer and matching glass jar
- Jar features an extra-wide hexagonal base for improved stability on the bench
- Made from durable soda-lime glass; 125 mm length
- Meets GG-U-681 requirements; replacement hydrometer available separately
Compliance
Complies with Federal Specification No. GG-U-681.
Specifications
Specifications
| Type | Urine Hydrometer |
| Accuracy | ±0.002 |
| Increments | 0.001 |
| Certifications/Compliance | No. GG-U-681 |
| Length (Metric) | 125 mm |
| Includes | Shot ballast weighted hydrometer and Glass jar |
| Material | Soda Lime Glass |
| For Use With (Application) | Urine analysis |