Learn More
Fisherbrand™ SureStain™ Stains
Catalog No. p-121871 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Wright Stain
Wright-Giemsa Stain
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
CS434D Wright-Giemsa Stain
CS432D Wright Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. CS434D
This item is not returnable. View return policy
Fast, reliable staining for differential blood cell analysis
  • For manual staining
  • Produce uniform results
  • Use for immersion or rack-type staining procedures
  • Entire procedure can be performed in 30 seconds
Order Info
32 oz. bottles.
Specifications
Quantity 32 oz.
For Use With (Application) For differential blood cell analysis
Product Type Wright-Giemsa Stain