Learn More
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring File Drawers
Catalog No. 13989120 Shop All Caplugs Products
Change view
Click to view available options
Color:
Dark Green
Catalog No. Color
13-989-120 Dark Green
1 options
Catalog No. 13-989-120 Supplier Fisher Scientific™ Supplier No. 2584200G10
Add to Cart
Edge
Add to Cart
Includes
A large I.D. label
Multiple stacking for maximum use of storage space
Multiple stacking for maximum use of storage space
  • Made of high-impact polystyrene
  • Removable inner sleeve drawer
Specifications
Product Type File Drawer
Color Dark Green
Holds 250 Embedding Rings or 400 Tissue Cassettes or a combination of both
Height (English) 2.25 in.
Height (Metric) 5.8 cm
Width (English) 9.5 in.
Width (Metric) 24.13 cm