Learn More
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets
Catalog No. 22025591
Change view
Click to view available options
Product Type:
Filing Cabinet
Catalog No. Product Type
22-025591 Filing Cabinet
1 options
Catalog No. 22-025591 Supplier National Scientific Supply Company, Inc Supplier No. HAFC606
Six-drawer model with six insert trays holds 900 embedding rings or 1800 tissue cassettes
Six-drawer model with six insert trays holds 900 embedding rings or 1800 tissue cassettes
  • For long-term, orderly storage, and easy retrieval of embedded cassettes or rings
  • Durable, tan metal cabinet
  • Available in six-drawer configuration for daily lab-bench usage
  • Clear plastic drawers feature removable, six-section insert trays with adjustable foam stops to keep rings or cassettes in a vertical orientation
Specifications
Product Type Filing Cabinet
Color Clear
Holds 900 Embedding Rings or 1800 Tissue Cassettes