Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Bench Conductivity Meter

Catalog No. 09330 Shop All Fisherbrand Products
Change view
Click to view available options
Calibration Points:
4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Points
09-330 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-330

missing translation for 'validMainframeMsg'

Traceable™ Bench Conductivity Meter measures conductivity, resistivity, dissolved solids, salinity, concentration, and temperature; includes certificate of traceability to standards provided by NIST™.

  • Features six modes — conductivity, resistivity, total dissolved solids, salinity, concentration and temperature readings
  • Ideal for ISO 9000 and government certification requirements
  • Autorange automatically selects most appropriate range for current reading unless feature is disabled
  • Controls allow operator to calibrate to solution standards; four calibration points ensure accuracy over entire range
Automatic or manual temperature compensation is defeatable
  • Automatic/manual temperature compensation with readings referenced to international standard of 25°C
  • Temperature compensation disable function fulfills USP-NF (United States Pharmacopeia, National Formulary, 645 Conductivity Measurement) requirements
Multiple applications plus connectivity
  • Measures conductivity in chromatography, biology, food, electronics and water analysis
  • Meets CAP, ASTM™, NCCLS, and ACS specifications in measuring reagent-grade water
  • Includes certificate to indicate instrument traceability to standards provided by the National Institute of Standards and Technology; calibrated by an ISO 17025 calibration laboratory accredited by A2LA
  • RS-232-C output allows readings to be downloaded to accessory datalogger or computer
Measurement Modes and Ranges
  • Conductivity Autoranging: 0.01 μS to 200 mS (in 5 ranges)
  • Resistance Autoranging: 0.001 to 20.00 megohms (in 2 ranges)
  • TDS Autoranging: 0.01 mg/L to 20.0 g/L (in 4 ranges)
  • Temperature: -30° to +130°C (-22° to +266°F)
  • Concentration: 0.000 to 9999 user units
  • Salinity: 2 to 42 ppt
  • Relative Accuracy: ±0.3%
Temperature Compensation
  • Types Automatic and manual, displays temperature °F/°C
  • Reference Temperature (conductivity): References to international standard 25°C (77°F)
Features
  • Electronic Calibration Supplied Internal (1000 °mhos and 0.001 megohm)
  • Data Output: RS-232-C, recorder jack
  • Display: 0.5 in. digit LCD
  • Net Weight: 1.5 lb. (0.7 kg)
  • Dimensions (L x W x H): 8.25 x 6 x 3.5 in. (21 x 15.2 x 8.9 cm)

Order Info

Replacement glass probe (09-326-2A) and unbreakable epoxy plastic probe (22-278143) are sold separately.

Specifications

Includes 115 V Power Adapter, 3 in. long (7.6 cm) Glass Probe, Receptacle to supply probe replatinizing current, 21 in. long (53.3 cm) Pivoting Probe Holder Arm, 9 V Battery
Accuracy (Conductivity) ±0.3%
Outputs RS232
For Use With (Application) Conductivity
Measurement Modes Conductivity, Resistivity, Dissolved Solid, Salinity, Concentration, Temperature
Calibration Points 4
Range (Conductivity) 0.01 μS to 200 mS
Range (Salinity) 2 to 42 ppt
Range (TDS) 0.01 mg/L to 20 g/L
Temperature Range (Metric) -30°C to +130°C
Temperature Range (English) -22°F to +266°F
Range (Resistivity) 0.001 to 2 megohm
Display Type LCD
Certifications/Compliance CAP, ASTM, NCCLS, ACS
Weight (Metric) 0.7 kg
Weight (English) 1.5 lb.
Detection Range 0.01 μS/cm to 200 mS/cm
Accuracy ±0.3%
Height (English) 8.25 in.
Height (Metric) 20.95 cm
Depth (Metric) 8.89 cm
Battery Type 9 V
Voltage 115 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.