Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Lollipop™ Shock/Waterproof Thermometer
SCI_ED

Catalog No. S04823 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +300°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S04823 -50°C to +300°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04823
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Offers minimum/maximum temperature readings. Fisherbrand Traceable Lollipop Shock/Waterproof Thermometer is water-resistant and is ideal for wet labs, wash-down areas, and all field applications.

  • Safer replacement of hazardous mercury units
  • Stainless-steel probe is resistant to most laboratory chemicals
  • Shockproof in drop tests up to 5 ft.
  • Memory recalls highest and lowest temperature readings which are updated every second
  • Solid-state construction eliminates dangers inherent in breakable glass
  • Operates without fail in all lab conditions - outdoors too
  • Bright and easy to view 0.25 in. high display
  • Traceable to NIST for accuracy
  • Temperature range: -50° to +300°C (-58° to +572°F)
  • Resolution: 0.1° from -20° to 200°; 1° outside this range
  • Accuracy: +/-1°C between 0° and 100°C
  • Stem diameter: 3.5mm (0.14 in.)
  • Stem length: 203mm (8 in.)
  • Overall length: 260mm (10.25 in.)
  • Width: 44.4mm (1.75 in.)
  • Depth: 12.7mm (0.5 in.)
  • Weight: 0.87 oz.

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Length (Metric) 33 cm
Resolution 0.1° from -20°C to 200°C
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Accuracy ±1°C
Display Digital
Width (Metric) 5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.