Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Refrigerator/Freezer Plus™ Thermometer

Catalog No. 06664269 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +70°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
06-664-269 -50°C to +70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-664-269
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

May be used for liquids, air/gas, semisolids, water baths and refrigerators

Monitors temperatures accurately in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Refigerator/Freezer Plus™ Thermometer includes sensor with 5mm diameter and 19mm length. May be used for liquids, air/gas, semisolids, water baths and refrigerators.

  • Minimum and maximum temperature memory
  • Triple display simultaneously shows minimum, maximum and current temperatures
  • High and low temperature alarm programmable in 1 degree increments
  • Includes calibration certificate

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.18 in.
Diameter (Metric) Probe 5 mm
Dimensions (L x W x H) 83 x 16 x 64 mm
For Use With (Application) Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
Includes Traceable certificate, AAA battery
Length (English) Cable 120 in.
Length (Metric) 83 mm
Length (Metric) Cable 304.8 cm
Material Plastic
Resolution 0.1°C
Weight (English) 4 oz.
Weight (Metric) 113.39 g
Width (English) 2.5 in.
Product Type Digital Thermometer
Length (English) 3.25 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Monitoring Temperature in Freezer
Accuracy ±0.5°C
Display Digital
Width (Metric) 6.35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.