Learn More
Fisherbrand™ Traceable™ Timer
SCI_ED
Catalog No. S94843 Shop All Fisherbrand Products
Change view
Click to view available options
Time Range:
99 hr. 59 min. 59 sec.
Catalog No. Time Range
S94843 99 hr. 59 min. 59 sec.
1 options
This item is not returnable. View return policy
Catalog No. S94843
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Easel bench stand, magnetic back, and battery
Four independent timing channels feature unique audible alarms for simultaneous timing
  • Unique memory returns channel to previously programmed setting at touch of a key
  • Counts up during alarming indicating elapsed time since alarming, 12/24 hour clock
Count up or down from 100 hours for longer duration experiments. Memory function returns the display to the programmed time.
Traceable™ Products have an individually-numbered Traceable™ Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Product Type Timer
Time Range 99 hr. 59 min. 59 sec.
Alarms Audible
Accuracy 0.0001
Resolution 1 sec.
Display LCD
Weight (English) 0.17 lb.
Weight (Metric) 0.08 kg
Includes Easel bench stand, magnetic back, and battery
Certifications/Compliance NIST