Learn More
Universal pH Indicator System Replacement Chart
Catalog No. I180 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
I180 Each
1 options
Catalog No. I180
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
  • Replacement Chart
  • As supplied with Universal pH indicator system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Universal pH indicator system
Includes 12 translucent color standards permanently mounted next to openings of the same size pH value of each color standard
Quantity Each
Type Indicator Solution Color Chart