Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Universal Swivel Clamp for Overhead Stirrers

Catalog No. 14512908 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Clamp
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-512-908 Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-512-908

missing translation for 'validMainframeMsg'

The universal swivel clamp lets you set the stirrer to any compound angle for best stirring action.

Made with precision machined aluminum rod for strength and durability

Has three knobs:

  • Knob one: Raises and lowers stirrer
  • Knob two: Locks stirrer on support rod
  • Knob three: Controls swivel setting

Specifications

For Use With (Equipment) Stirring stands from 12mm to 19mm dia.
Type Clamp
Material Cast Zinc, Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.