Learn More
Fisherbrand™ Vinyl Chuck Guard
Catalog No. 14500350 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Guard Accessory
Catalog No. Type
14-500-350 Guard Accessory
1 options
Catalog No. 14-500-350
For use with Fisherbrand Overhead Stirrers
  • Hole diameter will accommodates 5/16 or 3/8 in. diameter mixing shafts
  • Vinyl
  • Protects the chuck from splashes
  • Protects the user from the moving chuck
Specifications
For Use With (Equipment) Fisher Scientific™ Overhead Stirrer
For Use With (Application) Protects the chuck from splashes and protects the user from the moving chuck
Type Guard Accessory