Learn More
Fisherbrand™ MicroProbe™ Manual Staining System
Catalog No. 1518810 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Manual Staining System
Catalog No. Product Type
15-188-10 Manual Staining System
1 options
Catalog No. 15-188-10
Includes
MicroProbe Staining Station with eight reagent buckets and lids; Locator tray for Isolon holders; Four Isolon holders; Five 30-well rubber reagent isolators; Five 3-well rubber reagent isolators; Six absorbent pads; slide holder with 72 ProbeOn microscope slides; MicroProbe Incubator with lid, calibration block, and power cord; User manual; Application guide
Performs multistep staining protocols and simplifies standard chemical staining, immunostaining, and in situ hybridization
Compact system stains 20 slides at a time using capillary gap technology.
Staining Station
  • Eight reagent buckets with removable lids that can be filled to 20 or 100mL levels
  • Removable tray retains four plastic holders that accept Isolon™ reagent isolators or absorbent pads
Slide Holder Assembly and ProbeOn™ Microscope Slides
  • Allow users to move 20 ProbeOn slides (10 pairs) between staining station and incubator
  • Patented holder has lightweight, chemical-resistant plastic handle with numbered positions and indexing bump
Isolon™ Reagent Isolators
  • Available with three wells that contain milliliter volumes or with 30 smaller wells for microliter volumes of antibodies or DNA probes
  • Disposable, 30-well glass isolators are also available
MicroProbe Incubator
  • Heats from 30° to 110°C (86° to 230°F) in less than 10 minutes
  • Internal fan
  • Anodized aluminum incubation chamber
Compliance
CSA approved
Warranty and Services
One year
† Customer configurable for 100/220/240VAC. (Line cords must be ordered separately—contact your Fisher Scientific Customer Service Representative.)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Manual Staining System