Learn More
Fisherbrand™ 200 Series Safety Glasses
Catalog No. 191302089 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color
19-130-2089 Blue
19-130-2088 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-2089
You must be signed in and have a business account to purchase online.
Economical, lightweight protection
  • One-piece lens provides a full and unobstructed field of vision
  • Absorbs >99.9% of harmful UV light
  • Molded-in sideshields and browguards
  • Adjustable spatula temples
Certifications
CSA approved.
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Certifications/Compliance ANSI Z87.1+, ANSI Z80.3
Lens Coating Hardcoat
Shape Rectangle
Lens Tint Clear
Product Type Safety Glasses
Frame Color Blue
Temple Style Adjustable; spatula